VWR® Disposable Pasteur Pipettes, Glass
Supplier: VWR International
14673-010 14673-043 53283-916
14673-010CS 234.88 USD
14673-010 14673-043 53283-916
VWR® Disposable Pasteur Pipettes, Glass
Pipettes Pasteur/Transfer Pipettes
VWR® Disposable Pasteur Pipettes are manufactured from 3.3 expansion borosilicate glass, and feature fire-polished tops and a straight-cut tip.
  • Ideal for use in a wide range of laboratory applications
  • Fire-polished tips facilitate bulb attachment
  • 3.3 expansion borosilicate glass construction
Pasteur transfer pipettes are excellent for use in all general laboratory applications, including blood banks, hematology, chemistry, and bacteriology. Smooth, fire-polished tops facilitate bulb attachment. Glass pasteur pipettes have an inside diameter of 7 mm,More Product Information