Straight-Sided Jars, Clear, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W216903 W216904 W216905 W216906 W216907 W216908 W216909 W216910 W216911 W216912 W216913 W216914 W216919 W216920 W216921 W216922 W216923
14235-818CS 121.19 USD
14235-818 14235-820 14235-822 14235-824 14235-826 14235-828 14235-830 14235-832 14235-834 14235-836 14235-838 14235-840 14235-850 14235-852 14235-854 14235-856 14235-858
Straight-Sided Jars, Clear, Wheaton®, DWK Life Sciences
Jars General Use Jars
Wheaton Straight Sided jars are manufactured from clear soda lime glass that conforms to USP Type III requirements.
  • Jars are ideal for soil sampling and environmental applications
The wide mouth design with straight sides easily accommodates large solid samples, and allows for complete removal of contents.
A partitioned carton protects bottles from scratches during shipping, and becomes useful for storing samples. Each case of bottles also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required.