SP Wilmad-LabGlass Class B Graduated Cylinders with Standard Taper Stopper, TC, SP Industries
Supplier: SP Industries
LG-5530-106
14202-438EA 116.19 USD
14202-438 14203-594 14206-492 14208-574 14208-592 14209-224 14209-384 14210-736
SP Wilmad-LabGlass Class B Graduated Cylinders with Standard Taper Stopper, TC, SP Industries
Cylinders Measuring Cylinders
These cylinders are calibrated "to contain" (TC) at 20 °C.
  • Made of borosilicate glass
  • Cylinders feature white graduations and a Standard Taper stopper
  • Used to measure foam characteristics of emulsions and sedimentation values for solids
These laboratory, general-purpose grade graduated cylinders conform to ASTM E 1272 Style 2 Class B.