Anti-CD8b Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 53-5.8]

Supplier: Biolegend
140414
140414-BLEA 324.3 USD
140414-BL
Anti-CD8b Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 53-5.8]
Antibodies
Pacific Blue™ anti-mouse CD8b.2 [53-5.8]; Isotype: Rat IgG1, κ; Reactivity: mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

* Pacific Blue™ has a maximum emission of 455 nm when it is excited at 405 nm. Prior to using Pacific Blue™ conjugate for flow cytometric analysis, please verify your flowMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol CD8b
Environmentally Preferable
Antigen Synonyms Lyt-3|Ly-3|CD8B1
Antibody Type Primary
Clonality Monoclonal
Conjugation PB (Pacific Blue)
Clone 53-5.8
Reactivity Mouse
Host Rat
Gene ID 12526
Isotype IgG1 kappa
Immunogen Mouse thymus or spleen
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™.
Concentration 0.50 mg/mL
Storage Temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping Temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR