DECON-SPORE 200 Plus®, Veltek Associates
Supplier: Veltek Associates
DS200-05A DS200-04-1/2ZA DS200-04A DS200-06-16Z-01 DS200-03-13ZA DS200-06-16Z-02 DS200-02A DS200-07-16Z-01 DS200-01A DS200-03-2ZA DS200-03-1ZA DS200-05-1/2ZA
14003-710CS 369.58 USD
14003-710 14003-712 14003-708 14003-714 14220-126 14003-716 14220-122 14003-718 14220-120 14220-124 75795-622 76326-562 76326-564
DECON-SPORE 200 Plus®, Veltek Associates
Cleaning Agents Disinfectants
Designed for the sterilization of manufacturing, packaging, and filling equipment in aseptic processing areas.
  • Prefiltered to 0.2 um
  • Completely traceable and validated for sterility and shelf life
  • Effective disinfectant in water up to 400ppm hardness in the presence of 5% serum contamination and dried soap film residue
  • Avaliable in three configurations: concentrate, disinfectant use dilution, and sporicidal use dilution."
Certifications: Delivered with lot specific Certificate of Analysis and Certificate of Sterility.
Packaging: Double bagged in easy tear bag.
Order Now
SPECIFICATIONS
Environmentally Preferable