VWR® Low-Temperature Covers for Modular Heating Blocks
Supplier: VWR International
80863141 80863142 80863140
13259-303EA 176.36 USD
13259-303 13259-304 13259-301
VWR® Low-Temperature Covers for Modular Heating Blocks
Block Heaters Blocks
The Plexiglas® cover reduces air flow for additional temperature stability in low-temperature applications.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
Two sides open 6.4mm (1/4").
Order Now
SPECIFICATIONS
Environmentally Preferable