SP Bel-Art Vacuum Aspirator Pump, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F32947-0000
13197-066EA 27.64 USD
13197-066
SP Bel-Art Vacuum Aspirator Pump, Bel-Art Products, a part of SP
Pumps Drum/Transfer Pumps
High-density polyethylene aspirator pump operates on water pressure above 11psi (7584 N/m2).
  • HDPE construction
  • Maximum vacuum of 69.6 cm Hg
  • Corrosion-resistant check valve
The integral check valve resists corrosive filtrates and fumes. A quick-disconnect hose fitting accepts 6.4 to 9.5 mm (¼ to ⅜”) I.D. tubing. The male thread faucet connection is 9.5 mm (⅜”) I.D. Maximum attainable vacuum is 69.6 cm Hg (27.4”) with a flow of 6 liters (1.5 gallons) per minute.