Corning® BioCoat® Collagen I-Coated Flasks, Corning
Supplier: Corning
356484 354486 354484 356486 354487 354485 356485 356487
12777-072CS 522.56 USD
12777-072 12777-078 62405-025 12777-080 12777-082 12777-074 12777-076 12777-086
Corning® BioCoat® Collagen I-Coated Flasks, Corning
Flasks Cell Culture Flasks
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. In vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in
More Product Information