Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Supplier: QLA
GLA02K-01 GLA02K-DK GLA500-01 GLA500-DK GLA500-HR GLA900-01 GLA900-AD GLA900-AH GLA900-AV GLA900-CA GLA900-DK GLA900-EW GLA900-HR GLA900-ST GLA900-VK GLA9CR-AD GLA9CR-DK GLA9CR-ZM GLA9PK-01 PLA900-01 PLA900-AD PLA900-AH PLA900-AV PLA900-DK PLA900-HR GLA02K-HR GLA900-AST PLA900-PT PLA900-AP GLA900-AC GLA900-PT GLA9PK-DK GLA9PK-HR ASTM900-UN ASTM900-UNAM GLA9CR-AZM VESSTD-06H VESSTD-08H ASTM900-VK GLA9PK-AV GLA02K-EW GLA900-AEW GLA9PK-VK PLA900-EW GLA500-AV ASTM900-AUNP ASTM900-AVK ASTM900-UNP ASTM9PK-AUNP ASTM9PK-UNP GLA02K-AD GLA900-APT GLA900-AUNP GLA900-DKAP GLA900-DKP GLA900-UNP GLA9PK-AH GLA9PK-AUNP GLA9PK-UNP
11010-314EA 311 USD
11010-314 11010-316 11010-332 11010-334 11010-336 11010-338 11010-340 11010-342 11010-344 11010-346 11010-350 11010-352 11010-354 11010-356 11010-358 11010-362 11010-364 11010-366 11010-368 11010-404 11010-406 11010-408 11010-410 11010-412 11010-414 89039-830 89039-836 89039-842 89049-302 89080-324 89080-328 89080-330 89080-332 89132-946 89132-948 89133-036 89133-074 89133-076 89173-100 89173-122 93000-974 93000-976 93000-978 93000-982 75849-058 75849-060 75849-138 75849-140 75849-142 75849-148 75849-150 75849-208 75849-210 75849-212 75849-214 75849-216 75849-218 75849-224 75849-226 75849-228
Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Dissolution Apparatus
QLA manufactures a wide selection of Precision Molded and Standard Glass Vessels for most dissolution testers.
  • All glass vessels are made of borosilicate glass in the USA
  • Vessels are available in clear and amber
  • UltraCenter Precision, Standard and Apex Vessels available with both glass and plastic rims
Good dissolution vessels are critical in obtaining correct and consistent results
UltraCenter Precision Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting inMore Product Information