RTW Adjustable-Height Worktables, Production Basics
Supplier: Production Basics
3109 3063 3030 3005 3010 3100 3002 3105 3107 3114 3061 3000 3007 3011 3014 3015 3102 3110 3111 3115 3057 3001 3103 3050 3052 3055 3060 PB-3123
97008-308EA 993.08 USD
97008-308 97008-310 97008-312 11005-126 11005-130 11005-138 11005-124 11005-142 11005-144 11005-150 87000-320 11005-122 11005-128 11005-132 11005-134 11005-136 11005-140 11005-146 11005-148 11005-152 87000-316 87000-322 87000-324 87000-326 87000-310 87000-312 87000-314 87000-318 76239-040 76239-042 76239-044 76240-226 76240-228 76245-966 76245-968 76246-056 76246-236 76268-318
RTW Adjustable-Height Worktables, Production Basics
Furniture Tables and Workbenches
Ergonomic tables are constructed of 12-gauge steel and industrial-grade laminate with an abrasion-resistant finish.
  • Optional ESD laminate has static dissipative range of 106 to 109
  • Height is adjustable in 5 cm (2") increments from 76.2 to 91.4 cm (30 to 36")
Standard models include blue frame with white laminate work surface or gray ESD laminate work surface.
Load capacity: 750 lbs.
Ordering information: Tables include frame, work surface, four 5×5 cm (2×2") leg adjusters, leveling feet, and parts and hardware packet for assembly. ESD laminateMore Product Information