Passive Air Sampling Kits-Integrated (Stainless Steel and Siltek®-Treated), Restek
Supplier: Restek
22100 24160 24161 24167 24165 22101 24166 24168 24169 24216
10862-580EA 1311.99 USD
10862-580 10862-614 10862-616 10862-618 10862-738 10862-754 10862-966 10863-060 10863-062 10863-064 10863-098
Passive Air Sampling Kits-Integrated (Stainless Steel and Siltek®-Treated), Restek
Samplers Air Samplers
Restek’s passive air sampling kit incorporates all the hardware necessary to collect air samples and is easy to assemble for field sampling.
  • Provide accurate integrated sampling without a sampling pump
  • Siltek®-treated components ensure accurate sampling of active components
  • Excellent for sampling times from 0.5 hour to 125 hours
The improved filter design greatly reduces the number of potential leak sites.The passive air sampling kit is available in seven sampling flow ranges and in stainless steel or Siltek®-treated finish. The stainless steel kit isMore Product Information