Stomacher® 4500 Lab Blender Series, Seward
Supplier: Seward Laboratory System
30010500 30010550
10861-032EA 13745.13 USD
10861-032 10861-034
Stomacher® 4500 Lab Blender Series, Seward
Homogenizers Laboratory Paddle Blenders
The robust Stomacher® 4500 is ideal for the BAM large volume testing procedures from 1000 to 4500 ml.
  • Easy to clean
  • Easy bag loading
  • Programmable settings
Stomacher® lab blenders offer fast reliable blending and homogenisation of a broad range of laboratory samples. The Stomacher® machines work by the action of twin-reciprocating paddles on the sample contained inside a sterile plastic bag.
Expanding the food testing focused blenders, the Stomacher® 4500 BAM is the perfect tool for larger volume BAM testing procedures for extraction from 375 gMore Product Information