BT Lab Systems Shaking Incubator, G-Biosciences
Supplier: G-Biosciences
BT916 BT917
10856-422EA 3669.69 USD
10856-422 10856-526
BT Lab Systems Shaking Incubator, G-Biosciences
Shakers and Mixers Shaking Incubators
The Incubating Shaker is designed to heat, mix and shake multiple tube sizes and plates (See Related Products and Options to select required blocks)
  • Perfect radius and excellent dimensional technology blend with adjustable mixing speed
  • High efficient, programmable shaking and temperature control
  • Microprocessor controlled and accurate temperature and speed control
  • Various blocks for improved versatility (See Related Accessories)
  • Built-in temperature calibration
  • Short mixing function
  • DC brushless motor drive, long life, free of maintenance
  • Multiple safety
More Product Information