Septa for 2.0 ml Big Mouth Step Vials, Restek
Supplier: Restek
21148
10854-010PK 255.88 USD
10854-010
Septa for 2.0 ml Big Mouth Step Vials, Restek
Septa
These replacement septa are extremely economical accessories for use with big mouth step vials.
  • Screw-thread vial compatibility
  • Economical
  • Red PTFE/Silicone
Extra-wide openings decrease the number of broken needles with a larger target area. These septa necessarily help maintain a seal during autosampler use.