KleenGuard™ A40 Liquid and Particle Protection Boot Covers, Kimberly-Clark Professional
Supplier: Ansell Healthcare
36779
10853-852CS 1144.9 USD
10853-852
KleenGuard™ A40 Liquid and Particle Protection Boot Covers, Kimberly-Clark Professional
Disposable Garments
Keeping wearers comfortable while offering effective protective clothing is a must, and KleenGuard™ A40 liquid and particle protection boot covers are a great solution.
  • Designed to keep out debris, dry particulates, and liquid splashes
  • Elastic opening and elastic ankle
  • Serged seams
  • Highly durable PVC sole
  • Film laminate material offers better liquid and particulate barrier than Tyvek®
  • Passed NFPA 99 crtieria for antistatic materials
KleenGuard™ personal protective equipment (PPE) enables rather than inhibits, allowing for maximum productivity andMore Product Information