Kimtech™ A7 Ankle High Shoe Covers, Kimberly-Clark Professional
Supplier: Kimberly-Clark
47972 47973
10847-812CS 466.48 USD
10847-812 10847-818
Kimtech™ A7 Ankle High Shoe Covers, Kimberly-Clark Professional
Disposable Garments
Kimtech™ A7 ankle high shoe covers protect the shoelace area, closing a gap that could be a vehicle for contamination not addressed by traditional shoe covers.
  • Enhanced chemical and biological protection to help protect people and processes from liquid splashes
  • Passes ASTM F1670 for penetration of blood and body fluids
  • 30% more material is added to cover the ankle
  • Two elastic bands above and below the ankle for enhanced coverage and fit
  • Durable anti-skid soles for a improved grip
Kimtech™ A7 ankle high shoe covers provide foot protection that’s moreMore Product Information