DNAPac™ RP Column, Thermo Scientific
Supplier: Thermo Scientific
088919 088920 088921 088923 088924 088925 303324 950-00
10841-562EA 1292.5 USD
10841-562 10841-564 10841-566 10841-612 10841-614 10841-616 76420-872 TS950-00
DNAPac™ RP Column, Thermo Scientific
Chromatography Columns
The Thermo Scientific™ DNAPac™ RP is a reversed-phase (RP) liquid chromatography column designed for analysis of oligonucleotides and double-stranded (ds) DNA/RNA fragments using LC/UV or LC/MS.
  • For ion-pair reversed-phase (IP-RP) separations
  • High resolution and high throughput
  • Excellent MS compatibility
  • Wide operating pH range (0 to 14)
  • High temperature stability (up to 110 ºC)
DNAPac™ RP are designed for ion-pair reversed-phase (IP-RP) separations of both oligonucleotides and double-stranded nucleic acids. The unique column chemistry providesMore Product Information