OHM Metrics™ Wrist Strap Tester, Transforming Technologies
Supplier: Transforming Technologies
WST200
10837-052EA 160.7 USD
10837-052
OHM Metrics™ Wrist Strap Tester, Transforming Technologies
Static Control Monitors
The WST200 tester ensures that basic ESD protection is met by quickly and accurately measuring the resistance between an operator, wrist strap, and ground cord
With a test voltage of 9 volts DC, the unit is specially designed to test without harm to the operator. The WST200 verifies that a continuous grounding path between a wrist band and coil cord by indicating three levels of resistance; HI, LO, and OK. An easy to read LED light and an audible alarm provide measurement details. Test parameters are factory set to pass 800K–10M, but it is adjustable to match specific user requirements.
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable