Avantor Sterile PETG Single-Use Bottle Assemblies, Avantor Fluid Handling
Supplier: VWR International
10830-292 10830-294 10830-296 10830-298 10830-300 75835-988
10830-292CS 3362.69 USD
10830-292 10830-294 10830-296 10830-298 10830-300 75835-988
Avantor Sterile PETG Single-Use Bottle Assemblies, Avantor Fluid Handling
Bottles Cell Culture Bottles
Sterile PETG containers with weldable tubing provide the maximum amount of flexibility and reliability.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶
These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials.
When used with sterile Avantor tube sets, custom possibilities are endless.
Assemblies have been designed with a broad spectrum of applications in mind, includingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable