PREempt™ RTU Disinfectant Solutions, Contec
Supplier: Contec
21109 21101 21105
10822-454CS 166.01 USD
10822-454 10822-516 10822-518
PREempt™ RTU Disinfectant Solutions, Contec
Cleaning Agents Disinfectants
One-step disinfectant cleaner utilizing Accelerated Hydrogen Peroxide® (AHP).
  • Effective, fast-acting, one-step disinfectant cleaner
  • Convenient, ready-to-use disinfectant for daily use
  • Low residue and compatible with a wide range of materials
  • Odor-free for good operator acceptability
  • Emits no VOCs and does not require personal protection equipment (PPE)
  • Hydrogen peroxide breaks down into water and oxygen for easy disposal and minimal, to no environmental impact
  • Not classified as dangerous goods
PREempt™ RTU , a patented synergistic blend of commonlyMore Product Information