FiveGo™ F4 Portable DO Meters, METTLER TOLEDO®
Supplier: Mettler Toledo
30266884 30266885 30266886
10821-358EA 487.41 USD
10821-358 10821-360 10821-382
FiveGo™ F4 Portable DO Meters, METTLER TOLEDO®
Electrochemistry Meters Oxygen Meters
Ideally suited for mobile applications in the laboratory, at-line, or outdoors, the FiveGo™ portable instruments provide high quality dissolved oxygen measurements with the simple click of a button. The robust and waterproof design makes the FiveGo™ instruments the perfect companion to get reliable measurement data even in harsh conditions.
  • Intuitive operation - Measurement made easy
  • Ergonomic design - Comfortable handling
  • Foldable foot stand provides optimal viewing while situated on a lab bench
  • Waterproof - Ideal for wet conditions
  • IP 67 waterproof
More Product Information