Nystatin 10 mg/mL in methanol, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Fungicidin, Mycostatin

J67369-8EQ J67369-XF
10814-962PK 190.46 USD
10814-962 10814-964
Nystatin 10 mg/mL in methanol, sterile filtered
Nystatin

Liquid

Formula: C₄₇H₇₅NO₁₇
MW: 926.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25541651
CAS Number: 1400-61-9
EINECS: 215-749-0
UN: 1230
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR