Hettich ROTINA 380 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
380RBLOOD2 380RBLOOD2-BC 380RBLOOD4 380RBLOOD4-BC 380RBOTTLE 380RBOTTLE-BC 380RCELLCULT2 380RCELLCUL2BC 380RCELLCULT4 380BLOOD2-BC 380BLOOD4 380BLOOD4-BC 380BOTTLE 380BOTTLE-BC 380CELLCULT2 380CELLCULT2BC 380CELLCULT4 380CELLCULT4BC 380PLATE 380RCELLCUL4BC 380RPLATE 380BLOOD2 1701-01 1706-01
10812-254EA 21452.58 USD
10812-254 10812-256 10812-258 10812-260 10812-262 10812-264 10812-266 10812-268 10812-270 10812-372 10812-374 10812-376 10812-378 10812-380 10812-382 10812-384 10812-386 10812-388 10812-390 10812-392 10812-394 10813-206 75801-818 75801-820
Hettich ROTINA 380 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
The Hettich Lab Technology ROTINA 380 benchtop centrifuge series delivers versatility, capacity and performance in a compact footprint.
  • Max capacity – 4 x 290 mL
  • Max RPM/RCF – 15,000/24,400
  • Temperature control available
  • General purpose
  • Versa-Rotor (1 rotor for multiple applications)
  • Quiet operation – quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional design - easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety - self-closing, auto-locking lid with
More Product Information