UltraSOLV® ScrubTIPS® Cleaning Swabs with Grit Tips, Foamtec International
Supplier: Foamtec International
HT170060-5
10809-942CS 1699.83 USD
10809-942
UltraSOLV® ScrubTIPS® Cleaning Swabs with Grit Tips, Foamtec International
Swabs Swabs for Cleaning
These unique cleaning tools are ideal for polishing stains, scratches, and process residue from hard-to-clean surfaces such as electro-static chucks
Swabs are available with a dual tip or rigid tip and washed diamond or silicon carbide abrasives.
Ideal for removing flashing from sensitive medical devices and optics and maintaining high-precision injection molds.