Geobacillus stearothermophilus Spore Suspension, EMD Millipore
Supplier: MilliporeSigma
1.11499.0001
10809-762EA 322.79 USD
10809-762
Geobacillus stearothermophilus Spore Suspension, EMD Millipore
Microbiology Test Systems Antibiotic Tests
The Geobacillus stearothermophilus spore suspension is used in conjunction with the test agar for performing the antibiotics sulfonamide residue test according to KUNDRAT.
The test is based on agar diffusion, using spores of Geobacillus stearothermophilus as test organisms. The suspension should be kept in a refrigerator at +2 to +8 °C. At room temperature (up to +25 °C), this suspension can be kept for 1-2 days only. If kept in a refrigerator according to instructions, the test can be stored until the expiry date indicated. If used after the expiery date, the spores may begin to lose their activity.