Unistik® 3 Safety Lancets, Owen Mumford
Supplier: Owen Mumford
AT1022 AT1024 AT1028 AT1042 AT1047 AT1048 AT1002 AT1007 AT1008 AT1012 AT1062 AT1027
10804-085BX 31.92 USD
10804-085 10804-087 10804-091 10804-093 10804-097 10804-099 10804-101 10804-105 10804-107 10804-109 10804-115 76200-344
Unistik® 3 Safety Lancets, Owen Mumford
Blood Lancets
Unistik® 3 single-use, sterile safety lancets provide an easy and convenient way to test blood.
A simple one-click action activates the lancet, and the incorporated Comfort Zone Technology® (CZT) provides a comfortable blood sampling experience.
Gentle lancets are recommended for use in patient areas, such as pediatric and frequent testing application). Comfort lancets are suggested for use in blood glucose monitoring and HbA1C applications. Normal lancets are recommended for use in blood glucose monitoring, cholesterol, or HbA1C applications. Extra lancets are suggested for useMore Product Information