Miriad Rapid TP/HIV Antibody Test POU+, MedMira
Supplier: MEDMIRA
815311005992
10789-425CS 10373.25 USD
10789-425
Miriad Rapid TP/HIV Antibody Test POU+, MedMira
Clinical Diagnostic Supplies Clinical Reagents and Kits
Miriad Rapid TP/HIV Antibody Test POU+ employs MedMira rapid vertical flow technology to deliver the simultaneous diagnosis of Treponema pallidum bacteria (TP), the causitive agent of syphilis and HIV-1/2
Miriad Rapid TP/HIV Antibody Test POU+ features a built-in reagent control line, works with whole blood, serum and plasma specimens, and requires no additional equipment. Test kits can be stored for 12 months at 2–30°C (35.6–86°F).
In three easy steps, users can obtain multiple instant results on a single test cartridge.