Antibiotic Discs, Geno Bioscience
Supplier: G-Biosciences
D031-B D041-B D061-B D071-B D081-B
10781-872PK 61.68 USD
10781-872 10781-874 10781-876 10781-878 10781-880
Antibiotic Discs, Geno Bioscience
Microbiology Test Systems Antibiotic Tests
Antibiotic discs are used for screening and testing of antibiotic resistant gene from bacteria.
Small discs impregnated with different antibiotics (ampicillin, nalidixic acid, penicillin and tetracycline). Blank discs are available to prepare own screening discs.