Ingenio® Electroporation Kits and Solution, Mirus Bio
Supplier: Mirus Bio
MIR50111 MIR50112 MIR50113 MIR50114 MIR50115 MIR50116 MIR50117 MIR50118 MIR50119
10766-836EA 248.45 USD
10766-836 10766-838 10766-840 10766-842 10766-844 10766-846 10766-848 10766-850 10766-990
Ingenio® Electroporation Kits and Solution, Mirus Bio
Electroporation Systems Electroporation Kits
Ingenio® Electroporation Kits and Solution provide a universal, high efficiency, low toxicity solution for electroporation of DNA or siRNA into hard to transfect cell types.
  • High efficiency electroporation
  • Compatible with most conventional electroporation devices
  • Save money and reduce research costs without sacrificing performance
Ingenio® is provided a a standalone solution or as part of a kit with cuvettes (0.2 and 0.4 cm) and cell droppers. The 0.2 cm cuvettes are part of the Ingenio Electoproation Kit compatible with the Lonza-Amaxa® Nucleofector®More Product Information
Order Now
SPECIFICATIONS
Bioproduction Yes
Cell Specific Mammalian, Insect
Environmentally Preferable
CRISPR/Cas 9 Yes
DNA Yes
DNA and siRNA Yes
Electroporation Kits Yes
Insect Yes
miRNA Yes
mRNA Yes
Oligos Yes
Protein Delivery Yes
siRNA Yes