VWR® Disposable Plastic Transfer Pipette, Graduated
Supplier: VWR International
10754-264 10754-266 10754-268 10754-270 10754-272 10755-252
10754-264BX 323.31 USD
10754-264 10754-266 10754-268 10754-270 10754-272 10755-252
VWR® Disposable Plastic Transfer Pipette, Graduated
Pipettes Pasteur/Transfer Pipettes
VWR® Disposable Transfer Pipettes are designed to safely transfer and dispense liquids in all types of laboratories.
  • Pipettes are packaged using medical grade paper and plastic film
  • Non-toxic pipettes can be safely incinerated or heat-sealed and frozen at temperatures as low as –70 °C (–94 °F)
VWR® pipettes are produced using an optimal blend of low density polyethylene, allowing for highly controlled liquid dispensing.