ASTM Liquid-in-Glass Mercury Thermometers, Partial or Without Immersion, Thermco Products
Supplier: Thermco
ACC001C ACC001F ACC002C ACC002F ACC003C ACC003F ACC004C ACC004F ACC005C ACC005F ACC006C ACC006F ACC009C ACC009F ACC010C ACC010F ACC011C ACC011F ACC014C ACC014F ACC023C ACC024C ACC025C ACC027C ACC033C ACC033F ACC034C ACC034F ACC035C ACC035F ACC036C ACC037C ACC038C ACC049C ACC057C ACC057F ACC061C ACC061F ACC071C ACC071F ACC075F ACC082C ACC082F ACC083C ACC083F ACC084C ACC084F ACC096C ACC100C ACC101C ACC102C ACC103C ACC104C ACC105C ACC106C ACC107C ACC115C ACC119C ACC119F ACC133C ACC134C ACC135C ACC135F ACC137C ACC039C ACC040C ACC041C ACC042C ACC085C ACC085F ACC086C ACC086F ACC087C ACC087F ACC088C ACC088F ACC089C ACC090C ACC091C ACC092C ACC093C ACC094C ACC095C
10746-938EA 63.68 USD
10746-938 10746-940 10746-942 10746-944 10746-946 10746-948 10746-950 10746-952 10746-954 10746-956 10746-958 10746-960 10746-970 10746-972 10746-974 10746-976 10746-978 10746-980 10746-988 10746-990 10747-126 10747-128 10747-130 10747-134 10747-148 10747-150 10747-152 10747-154 10747-156 10747-158 10747-160 10747-162 10747-164 10747-168 10747-188 10747-190 10747-198 10747-200 10747-238 10747-240 10747-254 10747-268 10747-270 10747-272 10747-274 10747-276 10747-278 10747-280 10747-294 10747-296 10747-298 10747-300 10747-302 10747-304 10747-306 10747-308 10747-330 10747-340 10747-342 10747-376 10747-378 10747-380 10747-382 10747-388 10747-766 10747-768 10747-770 10747-772 10747-802 10747-804 10747-806 10747-808 10747-810 10747-812 10747-814 10747-816 10747-818 10747-820 10747-822 10747-824 10747-826 10747-828 10747-830
ASTM Liquid-in-Glass Mercury Thermometers, Partial or Without Immersion, Thermco Products
Thermometers Analog Thermometers
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil and petroleum testing.
These thermometers were designed for specific testing and can be calibrated to your specific requirements.
Ordering information: Accessories not included. Certificate of calibration not included. ASTM thermometers with a NIST TraceableMore Product Information