VWR® Traceable® Refrigerator/Freezer Ultra™ Thermometers
Supplier: VWR International
10500-584 10500-586 10500-588 10500-590 10500-592 10500-594 10500-596 75784-084 75784-086
10500-584EA 128.47 USD
10500-584 10500-586 10500-588 10500-590 10500-592 10500-594 10500-596 75784-084 75784-086
VWR® Traceable® Refrigerator/Freezer Ultra™ Thermometers
Thermometers Digital Thermometers
Triple display simultaneously shows minimum, maximum, and current temperatures for the specified probe.
  • Highly precise
  • Visual and audible alarm indicators
  • Alarm is user programmable
  • Designed to meet CDC requirements
Temperature range is -50 to 70°C (-58 to 158°F) with a resolution of 0.1° and an Ultra™ accuracy of ±0.5°C between -30 to 50°C (otherwise ±1°C)
When an out-of-range condition is sensed, the Smart-Alarm™ triggers. Smart-Alarm™ features visual and audible indicators that signal continuously until acknowledged, even if the parameterMore Product Information