Wide Mouth Glass Packer Bottles, Amber, C&G Containers
Supplier: C&G Containers
LAP002400110 LAP002900000 LAP004200010 LAP004200100 LAP004900002 LAP008200010 LAP008230010 LAP008900000 LAP016200010 LAP016900000 LAP032210010 LAP032900000 LAP042200010 LAP042900000 LAP083200010 LAP083900000 LAP002200002 LAP002308100 LAP016100000 LAP032100000 LAP032200000 LAP004100000 LAP083200000 LAP042200000 LAP008100010 LAP004100010
71000-634CS 946.58 USD
71000-634 71000-636 71000-638 71000-642 71000-644 71000-650 71000-652 71000-654 71000-656 71000-658 71000-660 71000-662 71000-664 71000-666 71000-668 71000-672 71000-674 71000-676 71000-678 71000-680 71000-682 71000-684 71001-632 71001-758 89137-856 89137-858 30617-854 30621-146 30621-148 30621-150 30621-152 30621-154 10496-900 10496-902
Wide Mouth Glass Packer Bottles, Amber, C&G Containers
Jars General Use Jars
These glass jars are used primarily for the collection of liquid media, soil, sediments, and slurry media for testing Semi-volatiles (Light Sensitive compounds), Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement for Pre-cleaned and Certified products. This certificate of analysis is included in eachMore Product Information