ig® Thymidine Auxotrophic EHA105ᵀʰʸ Agrobacterium Chemically Competent Cells, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
1304-05 1304-15
104090-482PK 386.54 USD
104090-482 104090-484
ig® Thymidine Auxotrophic EHA105ᵀʰʸ Agrobacterium Chemically Competent Cells, Intact Genomics
Competent Cells
Enables development of more efficient plant transformation systems.
  • Requires supplemental thymidine for growth
  • Reduced bacterial overgrowth during co-cultivation
  • Decreased need for antibiotics
Intact Genomics (ig®) Thymidine Auxotrophic EHA105Thy Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless 50 mg/L of thymidine is added to minimal medium.
This prevents the bacteria from overgrowing plant tissues when used for plant transformation. 
Thymidine-Auxotrophic EHA105Thy Agrobacterium
More Product Information