Single Channel Pipettes, Variable Volume
Supplier: Hach
BBP065 BBP078
104087-178EA 924.08 USD
104087-178 104087-180
Single Channel Pipettes, Variable Volume
Pipettes Micropipettes
These piston pipettes are handy, general-purpose microliter pipettes for transferring exact volumes of liquid. They operate on the principle of air expulsion, making use of disposable tips. The volume that is to be pipetted is shown in a clearly visible display on the pipette handle.
  • Fitted with mechanical tip ejector system to prevent possible contamination
  • Pipette is operated by means of the push button at the end of the handle
  • Factory calibrated; no recalibration is needed for routine use