Ionic Strength Adjustor for Ammonia
Supplier: Hach
2824349
104086-694EA 79.61 USD
104086-694
Ionic Strength Adjustor for Ammonia
Electrolytes Electrode Filling Solutions
Ammonia Ionic Strength Adjustor (ISA) buffer solution for ammonia determinations by electrode methods.