Gelex 2100 Secondary Standards Kits, Portable Turbidimeters
Supplier: Hach
2464105 2589200 2589000
97027-030EA 566.76 USD
97027-030 MSPP-2464105 104086-440 104086-436
Gelex 2100 Secondary Standards Kits, Portable Turbidimeters
Standards Turbidity Standards
Gelex Secondary Standards Kit, for 2100P and 2100Q Series Portable Turbidimeters (includes 1 sealed vial each, 0 to 10 NTU, 10 to 100 NTU, 100 to 1000 NTU).
Ordering information: Three per pack.