Sulfuric acid 0.035 N Volumetric

Supplier: Hach
Danger

2349732
97026-494EA 39.35 USD
97026-494 104086-128
Sulfuric acid 0.035 N Volumetric
Sulfuric acid

For alkalinity determination by drop-count titration test kit.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR