1-(2-Pyridylazo)-2-naphthol (PAN) solution_SC 0,1% Indicator

Supplier: Hach
Danger

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

2122426 2122432 2122449
97026-624EA 51.48 USD
97026-624 97026-644 97026-690 104085-844 104085-846 104085-848
1-(2-Pyridylazo)-2-naphthol (PAN) solution_SC 0,1% Indicator
1-(2-Pyridylazo)-2-naphthol

For determination of low level manganese by the PAN method


Hach method 8149. Range: 0.006 to 0.700 mg/L Mn.

Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Boiling Pt: 412
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Method 8149
Method Name PAN
Parameter Manganese
Platform Solution
Range 0.006 - 0.700 mg/L Mn


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR