Nessler reagent

Supplier: Hach

Synonyms: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution

2119432
104085-836PK 74.41 USD
104085-836
Nessler reagent
Nessler's reagent

The reagent is used for ammonia nitrogen determination in waste water by the Nessler method

MDL Number: MFCD00049656
CAS Number: 7783-33-7

Order Now

Specification Test Results

Range 0.02-2.50 mg/L NH3-N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR