ProCycle™ 0.2 ml 8-Strip PCR Tubes with Separate Flat Strip Caps
Supplier: Alkali Scientific
PC7064
104029-068EA 110.75 USD
104029-068
ProCycle™ 0.2 ml 8-Strip PCR Tubes with Separate Flat Strip Caps
Tubes PCR Tubes
Ensure secure sealing in PCR applications with Alkali Scientific™ ProCycle™ 0.2 ml 8-Strip PCR tubes featuring separate flat strip caps, available in packs of 1000.
  • 0.2 ml strip tubes with separate flat caps
  • Ultra-thin walls
  • Positive sealing
  • Fits standard spacing for 8-channel pipettes
  • Certified RNAse, DNAse and pyrogen free
Elevate your PCR experiments to new heights with the ProCycle™ 0.2 ml 8-Strip PCR tubes and separate flat strip caps. Crafted for rapid thermal transfer and robust sample protection, these tubes ensure precise sealing and reliableMore Product Information