Sodium dodecyl sulfate (SDS) 20%, CellPro™

Supplier: Alkali Scientific

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

PB0284
104028-852EA 109.22 USD
104028-852
Sodium dodecyl sulfate (SDS) 20%, CellPro™
Sodium dodecyl sulphate (SDS)

Alkali Scientific™ presents CellPro™ SDS, Sodium Dodecyl Sulfate, a high-quality reagent designed for protein denaturation and DNA/RNA extraction applications. With its exceptional purity and consistency, CellPro™ SDS ensures reliable results in molecular biology experiments, including Western blotting, DNA sequencing, and protein analysis. This versatile detergent effectively solubilizes hydrophobic molecules and disrupts non-covalent bonds, making it an essential tool for various laboratory protocols. Trust Alkali Scientific™ CellPro™ SDS for superior performance and reproducibility in your research endeavors.


  • Anionic wetting agent for both acid and alkaline solutions
  • For protein and lipid solubilization in SDS-PAGE protein analysis and nucleic acid purification
  • Prepared in 18 megaohm water and is 0.2μm filtered

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 3082
Merck Index: 14,08636

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR