Potassium chloride solution MMMA, CellPro™

Supplier: Alkali Scientific

Synonyms: Potassiumchloride

DS0193
104028-084EA 72.29 USD
104028-084
Potassium chloride solution MMMA, CellPro™
Potassium chloride

CellPro™ Potassium Chloride Solution stands as a trusted reagent in molecular biology research. Crafted to meticulous standards of purity and sterility, this solution offers researchers a reliable source of potassium ions for experimental procedures. With its sterile filtered formulation, CellPro™ Potassium Chloride Solution ensures consistent and reproducible results, empowering researchers in their molecular biology experiments.


  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
MDL Number: MFCD00011360
CAS Number: 7447-40-7
UN: 3248
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR