ProCycle™ 0.1 ml Low-Profile Rigid Stable 8-Strip PCR Tubes with Attached Individual Flat Caps
Supplier: Alkali Scientific
PC7065
104027-944EA 110.75 USD
104027-944
ProCycle™ 0.1 ml Low-Profile Rigid Stable 8-Strip PCR Tubes with Attached Individual Flat Caps
Tubes PCR Tubes
Experience stability in PCR reactions with Alkali Scientific™ ProCycle™ 0.1 ml low-profile rigid stable 8-Strip PCR tubes with attached individual flat caps.
  • Robust inter-well linkages make the strips rigid and easy to handle, preventing spills
  • Angularly-attached caps prevent lids and hinges from interfering with each other, allowing for easier operation in an SBS format rack
  • Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity
  • Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested
More Product Information