Upright Stainless Steel Freezer Rack for 2" Boxes, 5×4 Configuration
Supplier: Alkali Scientific
UFR-542
104027-232EA 112.29 USD
104027-232
Upright Stainless Steel Freezer Rack for 2" Boxes, 5×4 Configuration
Racks Cryogenic Box Racks
Maximize freezer space with upright stainless steel freezer rack for 2" boxes, available in a 5×4 configuration and accommodating up to 20 boxes.
  • Constructed of corrosion resistant stainless steel
  • Racks fit standard 2" cardboard, plastic or metal boxes
  • Security lock devices available for each rack
Optimize your freezer storage space with this upright stainless-steel freezer rack, designed for 2" boxes in a versatile 5×4 configuration. Offering a capacity of 20 boxes, this rack provides efficient organization and easy access to samples, streamliningMore Product Information