Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate

AC10280-0100
103508-400EA 201.88 USD
103508-400 103508-476
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate 98%
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) hydrate
Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MW: 551.14 g/mol (anhy)
Melting Pt: 188 …190 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00150755
CAS Number: 34369-07-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤10.0 % (K.F.)
Solubility (5 % in water) Clear colorless
HPLC ≥97.5 %
UV spectrophotometry molar absorptivity 15400 +/-2%, on dried substance
Ratio of Absorbances 0.13 to 0.17 (A280/A260)
Bacterial endotoxins (LAL-test) negative reaction (< 0.6 EU/mg)
Related substances ≤2.0 % (AMP + ADP) HPLC
pH 2.5 to 3.5 (5 % in water)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR