Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 1C6]

Supplier: Novus Biologicals
NBP2-46157
103398-644EA 706.28 USD
103398-644
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 1C6]
Antibodies
The CD30 / TNFRSF8 Antibody (1C6) from Novus Biologicals is a mouse monoclonal antibody to CD30 / TNFRSF8. This antibody reacts with human. The CD30 / TNFRSF8 Antibody (1C6) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR