Krazy Glue™ Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12646-05
103301-826CS 12.62 USD
103301-826
Krazy Glue™ Pen, Electron Microscopy Sciences
Stains Microscopy and staining reagents
The one glue we all know and have used.
  • Ready to use
  • Clear in color
  • Bonds immediately
Requires no mixing or preparation. This pen contains cyanoacrylate. Comes in a 3 g tube.
Order Now
SPECIFICATIONS
Environmentally Preferable