Anti-BLMH Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88640
103275-096EA 706.28 USD
103275-096
Anti-BLMH Rabbit Polyclonal Antibody
Antibodies
The BLMH / Bleomycin Hydrolase Antibody from Novus Biologicals is a rabbit polyclonal antibody to BLMH / Bleomycin Hydrolase. This antibody reacts with human, mouse, rat. The BLMH / Bleomycin Hydrolase Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: BLMH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR