Anti-IgA Goat Antibody

Supplier: Southern Biotechnology
2053-01
103259-992EA 241.92 USD
103259-992
Anti-IgA Goat Antibody
Antibodies
Purified Goat Anti-Human IgA antibody with minimal reactivity to mouse, bovine, and horse serum proteins for use in ELISA assays.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR